2,2-Dimethyl-1,3-dinitropropane

Supplier: Matrix Scientific

077537-500MG 077537-1G 077537-5G
102666-770EA 387.69 USD
102666-770 102666-772 102666-774
2,2-Dimethyl-1,3-dinitropropane
2,2-Dimethyl-1,3-dinitropropane

Hazard: Irritant

Formula: C₅H₁₀N₂O₄
MDL Number: MFCD00086898
CAS Number: 762-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR