3-Chloro-1,2,4-triazole

Supplier: Matrix Scientific

077534-500MG 077534-1G 077534-5G
102666-752EA 92.31 USD
102666-752 102666-754 102666-756
3-Chloro-1,2,4-triazole
3-Chloro-1,2,4-triazole

Hazard: Irritant

Formula: C₂H₂ClN₃
MW: 103.51 g/mol
MDL Number: MFCD00128812
CAS Number: 6818-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR