5-(2-Methoxyphenyl)-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

077528-500MG 077528-1G 077528-5G
102666-722EA 387.69 USD
102666-722 102666-724 102666-726
5-(2-Methoxyphenyl)-1,3,4-oxadiazol-2-amine
5-(2-Methoxyphenyl)-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₉H₉N₃O₂
MDL Number: MFCD01710631
CAS Number: 5711-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR