4-Amino-5-[(2-naphthyloxy)methyl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077511-500MG 077511-1G
102666-654EA 387.69 USD
102666-654 102666-656
4-Amino-5-[(2-naphthyloxy)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(2-naphthyloxy)methyl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₃H₁₂N₄OS
MDL Number: MFCD13370782
CAS Number: 4413-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR