4-Amino-5-propyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077508-500MG 077508-1G 077508-5G
102666-638EA 516.92 USD
102666-638 102666-640 102666-642
4-Amino-5-propyl-4H-1,2,4-triazole-3-thiol
4-Amino-5-propyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₅H₁₀N₄S
MDL Number: MFCD01806886
CAS Number: 30342-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR