4-Amino-5-(4-methylphenyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077504-500MG 077504-1G 077504-5G
102666-616EA 193.85 USD
102666-616 102666-618 102666-620
4-Amino-5-(4-methylphenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(4-methylphenyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₀N₄S
MDL Number: MFCD00834149
CAS Number: 13229-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR