4-Amino-5-(2-bromophenyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077503-500MG 077503-1G 077503-5G
102666-610EA 387.69 USD
102666-610 102666-612 102666-614
4-Amino-5-(2-bromophenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-bromophenyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₇BrN₄S
MDL Number: MFCD02046316
CAS Number: 61055-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR