4-Amino-5-(1H-benzimidazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077497-500MG 077497-1G
102666-582EA 516.92 USD
102666-582 102666-584
4-Amino-5-(1H-benzimidazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(1H-benzimidazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₀H₁₀N₆S
MDL Number: MFCD05882948
CAS Number: 791806-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR