4-Amino-5-(2-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077495-500MG 077495-1G 077495-5G
102666-570EA 387.69 USD
102666-570 102666-572 102666-574
4-Amino-5-(2-methoxyphenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₀N₄OS
MDL Number: MFCD01912738
CAS Number: 61019-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR