4-Amino-3-mercapto-6-methyl-1,2,4-triazin-5(4H)-one

Supplier: Matrix Scientific

077487-500MG 077487-1G 077487-5G
102666-522EA 387.69 USD
102666-522 102666-524 102666-526
4-Amino-3-mercapto-6-methyl-1,2,4-triazin-5(4H)-one
4-Amino-3-mercapto-6-methyl-1,2,4-triazin-5(4H)-one

Hazard: Irritant

Formula: C₄H₆N₄OS
MW: 158.18 g/mol
MDL Number: MFCD02660195
CAS Number: 22278-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR