4-Amino-5-(4-tert-butylphenyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077485-500MG 077485-1G 077485-5G
102666-510EA 387.69 USD
102666-510 102666-512 102666-514
4-Amino-5-(4-tert-butylphenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(4-tert-butylphenyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₂H₁₆N₄S
MDL Number: MFCD00665888
CAS Number: 349116-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR