4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077477-500MG
077477-1G
077477-5G
102666-462EA
387.69
USD
102666-462
102666-464
102666-466
4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol
4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₈H₈N₄S MW: 192.24 g/mol |
MDL Number:
MFCD00269264 CAS Number: 22706-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...