4-Amino-5-benzyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077476-500MG 077476-1G 077476-5G
102666-456EA 387.69 USD
102666-456 102666-458 102666-460
4-Amino-5-benzyl-4H-1,2,4-triazole-3-thiol
4-Amino-5-benzyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₀N₄S
MDL Number: MFCD00269274
CAS Number: 13373-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR