4-Amino-5-butyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077469-500MG 077469-1G 077469-5G
102666-422EA 387.69 USD
102666-422 102666-424 102666-426
4-Amino-5-butyl-4H-1,2,4-triazole-3-thiol
4-Amino-5-butyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₆H₁₂N₄S
MDL Number: MFCD02252170
CAS Number: 31821-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR