4-Amino-5-(phenoxymethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077467-500MG 077467-1G 077467-5G
102666-412EA 258.46 USD
102666-412 102666-414 102666-416
4-Amino-5-(phenoxymethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(phenoxymethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₀N₄OS
MDL Number: MFCD00269275
CAS Number: 4461-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR