4-Methyl-5,6,7,8-tetrahydroquinazolin-2-amine

Supplier: Matrix Scientific

077465-500MG 077465-1G 077465-5G
102666-400EA 258.46 USD
102666-400 102666-402 102666-404
4-Methyl-5,6,7,8-tetrahydroquinazolin-2-amine
4-Methyl-5,6,7,8-tetrahydroquinazolin-2-amine

Hazard: Irritant

Formula: C₉H₁₃N₃
MDL Number: MFCD06655499
CAS Number: 58544-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR