5-Propylisoxazole-4-carboxylic acid

Supplier: Matrix Scientific

077464-500MG 077464-1G 077464-5G
102666-394EA 387.69 USD
102666-394 102666-396 102666-398
5-Propylisoxazole-4-carboxylic acid
5-Propylisoxazole-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD09756503
CAS Number: 134541-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR