2-Amino-4-phenylpyrimidine-5-carboxylic acid

Supplier: Matrix Scientific

077463-500MG 077463-1G 077463-5G
102666-388EA 193.85 USD
102666-388 102666-390 102666-392
2-Amino-4-phenylpyrimidine-5-carboxylic acid
2-Amino-4-phenylpyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉N₃O₂
MDL Number: MFCD08669646
CAS Number: 91093-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR