1-Ethyl-1H-pyrazole-5-carbaldehyde
Supplier: Matrix Scientific
077460-1G
077460-5G
102666-372EA
269.23
USD
102666-372
102666-374
1-Ethyl-1H-pyrazole-5-carbaldehyde
1-Ethyl-1H-pyrazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₆H₈N₂O MW: 124.14 g/mol |
MDL Number:
MFCD03130036 CAS Number: 902837-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...