3-(1-Methyl-1H-pyrazol-4-yl)propan-1-ol
Supplier: Matrix Scientific
077459-500MG
077459-1G
077459-5G
102666-364EA
258.46
USD
102666-364
102666-366
102666-368
3-(1-Methyl-1H-pyrazol-4-yl)propan-1-ol
3-(1-Methyl-1H-pyrazol-4-yl)propan-1-ol
Hazard: Irritant
Formula:
C₇H₁₂N₂O |
MDL Number:
MFCD07440134 CAS Number: 192661-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...