3-(1H-Pyrazol-4-yl)propan-1-ol

Supplier: Matrix Scientific

077458-500MG 077458-1G 077458-5G
102666-358EA 646.15 USD
102666-358 102666-360 102666-362
3-(1H-Pyrazol-4-yl)propan-1-ol
3-(1H-Pyrazol-4-yl)propan-1-ol

Hazard: Irritant

Formula: C₆H₁₀N₂O
MDL Number: MFCD01823396
CAS Number: 60951-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR