1-Pyrazin-2-ylacetone

Supplier: Matrix Scientific

077453-500MG 077453-1G 077453-5G
102666-328EA 775.38 USD
102666-328 102666-330 102666-332
1-Pyrazin-2-ylacetone
1-Pyrazin-2-ylacetone

Hazard: Irritant

Formula: C₇H₈N₂O
MDL Number: MFCD08572186
CAS Number: 6784-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR