[4-(1H-Pyrazol-5-yl)phenyl]boronic acid

Supplier: Matrix Scientific

077452-500MG 077452-1G 077452-5G
102666-322EA 775.38 USD
102666-322 102666-324 102666-326
[4-(1H-Pyrazol-5-yl)phenyl]boronic acid
[4-(1H-Pyrazol-5-yl)phenyl]boronic acid

Hazard: Irritant

Formula: C₉H₉BN₂O₂
MDL Number: MFCD06739074
CAS Number: 1174304-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR