Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 5-amino-1,3,4-oxadiazole-2-carboxylate

077450-500MG 077450-1G
102666-312EA 193.85 USD
102666-312 102666-314
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate
Ethyl-5-amino-1,3,4-oxadiazole-2-carboxylate

Hazard: Irritant

Formula: C₅H₇N₃O₃
MW: 157.13 g/mol
MDL Number: MFCD03425200
CAS Number: 4970-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR