5-Amino-1,3,4-oxadiazole-2-carboxylic acid

Supplier: Matrix Scientific

077449-500MG 077449-1G
102666-308EA 621.54 USD
102666-308 102666-310
5-Amino-1,3,4-oxadiazole-2-carboxylic acid
5-Amino-1,3,4-oxadiazole-2-carboxylic acid

Hazard: Irritant

Formula: C₃H₃N₃O₃
MDL Number: MFCD19203574
CAS Number: 4970-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR