2-Methyl-1H-pyrrole-3-carboxylic acid

Supplier: Matrix Scientific

077448-500MG 077448-1G 077448-5G
102666-302EA 492.31 USD
102666-302 102666-304 102666-306
2-Methyl-1H-pyrrole-3-carboxylic acid
2-Methyl-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₇NO₂
MW: 125.13 g/mol
MDL Number: MFCD01625949
CAS Number: 37102-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR