tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate

Supplier: Matrix Scientific

Synonyms: 4-(Bromoacetyl)-1-Boc-piperazine, tert-Butyl 4-(2-bromoacetyl)piperazine-1-carboxylate

077445-500MG 077445-1G 077445-5G
102666-288EA 269.23 USD
102666-288 102666-290 102666-292
tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate
tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉BrN₂O₃
MW: 307.19 g/mol
MDL Number: MFCD16990579
CAS Number: 112257-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR