Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-amino-1-methyl-1H-pyrazole-4-carboxylate

077444-500MG 077444-1G
102666-284EA 40 USD
102666-284 102666-286
Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate
Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
MDL Number: MFCD00101268
CAS Number: 21230-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR