2,4-Dichloro-6-ethyl-1,3,5-triazine

Supplier: Matrix Scientific

077441-500MG 077441-1G 077441-5G
102666-268EA 387.69 USD
102666-268 102666-270 102666-272
2,4-Dichloro-6-ethyl-1,3,5-triazine
2,4-Dichloro-6-ethyl-1,3,5-triazine

Hazard: Irritant

Formula: C₅H₅Cl₂N₃
MDL Number: MFCD15203623
CAS Number: 698-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR