N-Methyl-1-pyridin-4-ylethanamine

Supplier: Matrix Scientific

077438-500MG 077438-1G 077438-5G
102666-250EA 193.85 USD
102666-250 102666-252 102666-254
N-Methyl-1-pyridin-4-ylethanamine
N-Methyl-1-pyridin-4-ylethanamine

Hazard: Irritant

Formula: C₈H₁₂N₂
MDL Number: MFCD09048938
CAS Number: 158958-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR