2-(3-Nitro-1H-pyrazol-1-yl)acetamide

Supplier: Matrix Scientific

077426-500MG 077426-1G 077426-5G
102666-180EA 258.46 USD
102666-180 102666-182 102666-184
2-(3-Nitro-1H-pyrazol-1-yl)acetamide
2-(3-Nitro-1H-pyrazol-1-yl)acetamide

Hazard: Irritant

Formula: C₅H₆N₄O₃
MDL Number: MFCD01622830
CAS Number: 1003011-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR