5-Ethyl-1-methyl-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

077424-500MG 077424-1G 077424-5G
102666-168EA 516.92 USD
102666-168 102666-170 102666-172
5-Ethyl-1-methyl-1H-pyrazole-3-carboxylic acid
5-Ethyl-1-methyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD04115075
CAS Number: 165744-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR