1,5-Dimethyl-1H-pyrazole-4-carboxamide
Supplier: Matrix Scientific
077419-500MG
077419-1G
077419-5G
102666-138EA
516.92
USD
102666-138
102666-140
102666-142
1,5-Dimethyl-1H-pyrazole-4-carboxamide
1,5-Dimethyl-1H-pyrazole-4-carboxamide
Hazard: Irritant
Formula:
C₆H₉N₃O |
MDL Number:
MFCD11643973 CAS Number: 103589-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...