1-Methyl-3-nitro-1H-pyrazole-5-carboxylic acid

Supplier: Matrix Scientific

077417-500MG 077417-1G
102666-128EA 453.85 USD
102666-128 102666-130
1-Methyl-3-nitro-1H-pyrazole-5-carboxylic acid
1-Methyl-3-nitro-1H-pyrazole-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₅N₃O₄
MDL Number: MFCD06805246
CAS Number: 177409-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR