2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)acetamide

Supplier: Matrix Scientific

077416-500MG 077416-1G
102666-124EA 387.69 USD
102666-124 102666-126
2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)acetamide
2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)acetamide

Hazard: Irritant

Formula: C₈H₁₃N₃O
MDL Number: MFCD08685646
CAS Number: 1017502-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR