5-[(2-Nitrophenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077407-500MG 077407-1G 077407-5G
102666-078EA 387.69 USD
102666-078 102666-080 102666-082
5-[(2-Nitrophenoxy)methyl]-2-furoic acid
5-[(2-Nitrophenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₂H₉NO₆
MW: 263.21 g/mol
MDL Number: MFCD02090863
CAS Number: 364621-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR