5-[(2-Bromophenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077402-500MG 077402-1G 077402-5G
102666-048EA 387.69 USD
102666-048 102666-050 102666-052
5-[(2-Bromophenoxy)methyl]-2-furoic acid
5-[(2-Bromophenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₂H₉BrO₄
MDL Number: MFCD01858564
CAS Number: 364739-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR