1-(2,3-Dimethylphenyl)-1H-pyrrole-2,5-dione

Supplier: Matrix Scientific

077389-500MG 077389-1G 077389-5G
102665-972EA 193.85 USD
102665-972 102665-974 102665-976
1-(2,3-Dimethylphenyl)-1H-pyrrole-2,5-dione
1-(2,3-Dimethylphenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₂H₁₁NO₂
MDL Number: MFCD00030660
CAS Number: 31581-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR