Methyl-5-methylthiophene-2-carboxylate
Supplier: Matrix Scientific
Synonyms:
Methyl 5-methylthiophene-2-carboxylate
077383-500MG
077383-1G
077383-5G
102665-936EA
107.19
USD
102665-936
102665-938
102665-940
Methyl-5-methylthiophene-2-carboxylate
Methyl-5-methylthiophene-2-carboxylate
Hazard: Irritant
Formula:
C₇H₈O₂S MW: 156.2 g/mol |
MDL Number:
MFCD06203669 CAS Number: 19432-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...