6-Methylthieno[2,3-d]pyrimidin-4(3H)-one

Supplier: Matrix Scientific

Synonyms: 6-Methyl-3H-thieno[2,3-d]pyrimidin-4-one, 6-Methylthieno[2,3-d]pyrimidin-4-ol

077382-500MG 077382-1G 077382-5G
102665-930EA 258.46 USD
102665-930 102665-932 102665-934
6-Methylthieno[2,3-d]pyrimidin-4(3H)-one
6-Methylthieno[2,3-d]pyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₇H₆N₂OS
MW: 166.2 g/mol
MDL Number: MFCD01570294
CAS Number: 108831-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR