Methyl-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 5-methylthiophene-3-carboxylate

077380-500MG 077380-1G 077380-5G
102665-918EA 461.54 USD
102665-918 102665-920 102665-922
Methyl-5-methylthiophene-3-carboxylate
Methyl-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₇H₈O₂S
MDL Number: MFCD02245126
CAS Number: 88770-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR