2-Amino-5-methylthiophene-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-methyl-3-thiophenecarboxylic acid

077379-500MG 077379-1G
102665-914EA 516.92 USD
102665-914 102665-916
2-Amino-5-methylthiophene-3-carboxylic acid
2-Amino-5-methylthiophene-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₇NO₂S
MDL Number: MFCD05664138
CAS Number: 41940-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR