Methyl-5-methyl-1H-1,2,3-triazole-4-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 5-methyl-1H-1,2,3-triazole-4-carboxylate

077378-500MG 077378-1G 077378-5G
102665-908EA 646.15 USD
102665-908 102665-910 102665-912
Methyl-5-methyl-1H-1,2,3-triazole-4-carboxylate
Methyl-5-methyl-1H-1,2,3-triazole-4-carboxylate

Hazard: Irritant

Formula: C₅H₇N₃O₂
MDL Number: MFCD18381938
CAS Number: 60419-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR