3-(Ethoxymethylene)pentane-2,4-dione

Supplier: Matrix Scientific

077371-500MG 077371-1G 077371-5G
102665-866EA 161.54 USD
102665-866 102665-868 102665-870
3-(Ethoxymethylene)pentane-2,4-dione
3-(Ethoxymethylene)pentane-2,4-dione

Hazard: Irritant

Formula: C₈H₁₂O₃
MDL Number: MFCD07368652
CAS Number: 33884-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR