4,4,5,5-Tetrachloro-1,3-dioxolan-2-one

Supplier: Matrix Scientific

077366-500MG 077366-1G 077366-5G
102665-836EA 184.62 USD
102665-836 102665-838 102665-840
4,4,5,5-Tetrachloro-1,3-dioxolan-2-one
4,4,5,5-Tetrachloro-1,3-dioxolan-2-one

Hazard: Irritant

Formula: C₃Cl₄O₃
MDL Number: MFCD00030631
CAS Number: 22432-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR