2,5-Dibromofuran

Supplier: Matrix Scientific

077362-500MG 077362-1G 077362-5G
102665-812EA 387.69 USD
102665-812 102665-814 102665-816
2,5-Dibromofuran
2,5-Dibromofuran

Hazard: Irritant

Formula: C₄H₂Br₂O
MDL Number: MFCD00191342
CAS Number: 32460-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR