5-[(4-Chlorophenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077355-500MG 077355-1G 077355-5G
102665-770EA 387.69 USD
102665-770 102665-772 102665-774
5-[(4-Chlorophenoxy)methyl]-2-furoic acid
5-[(4-Chlorophenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₂H₉ClO₄
MDL Number: MFCD02253883
CAS Number: 74556-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR