Methyl-4-cyclopropyl-2,4-dioxobutanoate

Supplier: Matrix Scientific

077354-500MG 077354-1G 077354-5G
102665-764EA 387.69 USD
102665-764 102665-766 102665-768
Methyl-4-cyclopropyl-2,4-dioxobutanoate
Methyl-4-cyclopropyl-2,4-dioxobutanoate

Hazard: Irritant

Formula: C₈H₁₀O₄
MDL Number: MFCD04967381
CAS Number: 167408-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR