Methyl-2-methoxy-6-methylbenzoate

Supplier: Matrix Scientific

077350-500MG 077350-1G 077350-5G
102665-742EA 387.69 USD
102665-742 102665-744 102665-746
Methyl-2-methoxy-6-methylbenzoate
Methyl-2-methoxy-6-methylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MDL Number: MFCD08235005
CAS Number: 79383-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR