Methyl-3-(chloromethyl)-4-methoxybenzoate

Supplier: Matrix Scientific

077349-500MG 077349-1G 077349-5G
102665-736EA 775.38 USD
102665-736 102665-738 102665-740
Methyl-3-(chloromethyl)-4-methoxybenzoate
Methyl-3-(chloromethyl)-4-methoxybenzoate

Hazard: Irritant

Formula: C₁₀H₁₁ClO₃
MDL Number: MFCD05864592
CAS Number: 36755-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR